COMH04238B56 Combi-Blocks, Inc.

1-(3,5-Dimethyl-1h-pyrazol-1-yl)-2-methylpropan-2-ol

  • MDL number MFCD12802354
  • Cas Number : 527677-31-2
  • Molecular Weight : 168.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6898

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6898
Availability details for
Quantity
  • Availability
  • Add to Cart