COMH04238B6C Combi-Blocks, Inc.

6-Isopropyl-2-pyridin-2-ylquinoline-4-carboxylic acid

  • MDL number MFCD05175923
  • Cas Number : 897561-16-9
  • Molecular Weight : 292.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6920

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6920
Availability details for
Quantity
  • Availability
  • Add to Cart