COMH04238BA8 Combi-Blocks, Inc.

6-Bromo-7-chloro-8-methyl-2-phenylquinoline-4-carboxylic acid

  • MDL number MFCD06127498
  • Cas Number : 894552-27-3
  • Molecular Weight : 376.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6980

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6980
Availability details for
Quantity
  • Availability
  • Add to Cart