COMH04238BB9 Combi-Blocks, Inc.

Methyl n-(3,5-dichlorophenyl)-n-(phenylsulfonyl)glycinate

  • MDL number MFCD02218808
  • Cas Number : 425605-85-2
  • Molecular Weight : 374.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6997

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6997
Availability details for
Quantity
  • Availability
  • Add to Cart