COMH04238BD2 Combi-Blocks, Inc.

5-[(2-Iodophenoxy)methyl]-4-phenyl-4h-1,2,4-triazole-3-thiol

  • MDL number MFCD01809921
  • Cas Number : 345989-01-7
  • Molecular Weight : 409.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7024

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7024
Availability details for
Quantity
  • Availability
  • Add to Cart