COMH04238BDB Combi-Blocks, Inc.

6-Chloro-2-(4-ethylphenyl)quinoline-4-carboxylic acid

  • MDL number MFCD03420103
  • Cas Number : 897559-96-5
  • Molecular Weight : 311.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7033

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7033
Availability details for
Quantity
  • Availability
  • Add to Cart