COMH04238BEB Combi-Blocks, Inc.

3-Nitro-n-(2-phenoxyethyl)benzamide

  • MDL number MFCD05611305
  • Cas Number : 903796-25-8
  • Molecular Weight : 286.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7049

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7049
Availability details for
Quantity
  • Availability
  • Add to Cart