COMH04238BEC Combi-Blocks, Inc.

[3-(4-Fluorophenyl)-5,9-dimethyl-7-oxo-7h-furo[3,2-g]chromen-6-yl]acetic acid

  • MDL number MFCD03661809
  • Cas Number : 664366-14-7
  • Molecular Weight : 366.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7050

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7050
Availability details for
Quantity
  • Availability
  • Add to Cart