COMH04238C0A Combi-Blocks, Inc.

2-(3,5-Dimethyl-1h-pyrazol-1-yl)propanoic acid

  • MDL number MFCD03170175
  • Cas Number : 956508-33-1
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7080

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7080
Availability details for
Quantity
  • Availability
  • Add to Cart