COMH04238C77 Combi-Blocks, Inc.

2-Chloro-n-(2-iodophenyl)acetamide

  • MDL number MFCD02580942
  • Cas Number : 66947-88-4
  • Molecular Weight : 295.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7190

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7190
Availability details for
Quantity
  • Availability
  • Add to Cart