COMH04238CE0 Combi-Blocks, Inc.

Isopropyl 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta[b]thiophene-3-carboxylate

  • MDL number MFCD01114971
  • Cas Number : 351983-32-9
  • Molecular Weight : 253.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7297

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7297
Availability details for
Quantity
  • Availability
  • Add to Cart