COMH04238D64 Combi-Blocks, Inc.

6-Bromo-2-(3-methoxyphenyl)quinoline-4-carboxylic acid

  • MDL number MFCD00845856
  • Cas Number : 351329-64-1
  • Molecular Weight : 358.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7432

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7432
Availability details for
Quantity
  • Availability
  • Add to Cart