COMH04238D67 Combi-Blocks, Inc.

(2-Aminophenyl)[bis(2-methylphenyl)]methanol

  • MDL number MFCD05857301
  • Cas Number : 94964-71-3
  • Molecular Weight : 303.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7435

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7435
Availability details for
Quantity
  • Availability
  • Add to Cart