COMH04238D6B Combi-Blocks, Inc.

Methyl 2-[(2-methoxy-2-oxoethyl)thio]benzoate

  • MDL number MFCD03855443
  • Cas Number : 6287-88-3
  • Molecular Weight : 240.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7439

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7439
Availability details for
Quantity
  • Availability
  • Add to Cart