COMH04238D91 Combi-Blocks, Inc.

(2Z)-2-(3-Fluorobenzylidene)-6-hydroxy-1-benzofuran-3(2h)-one

  • MDL number MFCD04158995
  • Cas Number : 620545-88-2
  • Molecular Weight : 256.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7477

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7477
Availability details for
Quantity
  • Availability
  • Add to Cart