COMH04238D9B Combi-Blocks, Inc.

Methyl (7-hydroxy-8-methyl-2-oxo-2h-chromen-4-yl)acetate

  • MDL number MFCD06001867
  • Cas Number : 714206-31-2
  • Molecular Weight : 248.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7488

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7488
Availability details for
Quantity
  • Availability
  • Add to Cart