COMH04238E08 Combi-Blocks, Inc.

2-Butoxy-5-chlorobenzoyl chloride

  • MDL number MFCD11941221
  • Cas Number : 62910-45-6
  • Molecular Weight : 247.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7600

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7600
Availability details for
Quantity
  • Availability
  • Add to Cart