COMH04238E18 Combi-Blocks, Inc.

Ethyl 4-methyl-2-pyrrolidin-1-yl-1,3-thiazole-5-carboxylate

  • MDL number MFCD14756688
  • Cas Number : 950269-69-9
  • Molecular Weight : 240.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7616

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7616
Availability details for
Quantity
  • Availability
  • Add to Cart