COMH04238E33 Combi-Blocks, Inc.

2-Amino-6-ethyl-n-(4-methylphenyl)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide

  • MDL number MFCD03422604
  • Cas Number : 667412-54-6
  • Molecular Weight : 314.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7643

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7643
Availability details for
Quantity
  • Availability
  • Add to Cart