COMH04238E5F Combi-Blocks, Inc.

N-1-Adamantyl-n-ethylamine hydrochloride

  • MDL number MFCD01674015
  • Cas Number : 3717-44-0
  • Molecular Weight : 215.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7687

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7687
Availability details for
Quantity
  • Availability
  • Add to Cart