COMH04238E81 Combi-Blocks, Inc.

Methyl 3-hydroxy-4-nitro-2-naphthoate

  • MDL number MFCD02615337
  • Cas Number : 5394-81-0
  • Molecular Weight : 247.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7721

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7721
Availability details for
Quantity
  • Availability
  • Add to Cart