COMH04238E93 Combi-Blocks, Inc.

4-Allyl-5-(2-phenylquinolin-4-yl)-4h-1,2,4-triazole-3-thiol

  • MDL number MFCD03423482
  • Cas Number : 847503-25-7
  • Molecular Weight : 344.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7740

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7740
Availability details for
Quantity
  • Availability
  • Add to Cart