COMH04238EA0 Combi-Blocks, Inc.

2-[(4-Formyl-2-methoxyphenoxy)methyl]benzonitrile

  • MDL number MFCD02629360
  • Cas Number : 443289-08-5
  • Molecular Weight : 267.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7753

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7753
Availability details for
Quantity
  • Availability
  • Add to Cart