COMH04238EBA Combi-Blocks, Inc.

1-(2-Chlorobenzyl)-3-methyl-1h-pyrazol-5-amine hydrochloride

  • MDL number MFCD13250082
  • Cas Number : 3524-40-1
  • Molecular Weight : 258.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7779

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7779
Availability details for
Quantity
  • Availability
  • Add to Cart