COMH04238EC8 Combi-Blocks, Inc.

N-(4-Bromophenyl)-4-hydrazino-4-oxobutanamide

  • MDL number MFCD02255632
  • Cas Number : 443863-36-3
  • Molecular Weight : 286.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7793

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7793
Availability details for
Quantity
  • Availability
  • Add to Cart