COMH04238EE8 Combi-Blocks, Inc.

6-tert-Butyl-1-benzothiophene-3-carbohydrazide

  • MDL number MFCD03419867
  • Cas Number : 956576-47-9
  • Molecular Weight : 248.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7825

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7825
Availability details for
Quantity
  • Availability
  • Add to Cart