COMH04238EFB Combi-Blocks, Inc.

1-Propylpiperazin-2-one

  • MDL number MFCD14707896
  • Cas Number : 65464-10-0
  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7845

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7845
Availability details for
Quantity
  • Availability
  • Add to Cart