COMH04238F18 Combi-Blocks, Inc.

2-(1,3-Dioxooctahydro-2h-4,7-methanoisoindol-2-yl)-3-phenylpropanoic acid

  • MDL number MFCD12197718
  • Cas Number : 2173071-88-8
  • Molecular Weight : 313.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7876

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7876
Availability details for
Quantity
  • Availability
  • Add to Cart