COMH04238F1B Combi-Blocks, Inc.

3-Chloro-n-(3,5-dichlorophenyl)propanamide

  • MDL number MFCD02767295
  • Cas Number : 90348-83-7
  • Molecular Weight : 252.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7879

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7879
Availability details for
Quantity
  • Availability
  • Add to Cart