COMH04238F1E Combi-Blocks, Inc.

1-(1-Adamantyl)-4,4,4-trifluorobutane-1,3-dione

  • MDL number MFCD03419461
  • Cas Number : 758709-48-7
  • Molecular Weight : 274.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7882

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7882
Availability details for
Quantity
  • Availability
  • Add to Cart