COMH04238F58 Combi-Blocks, Inc.

1-([(5-Methyl-1,3-thiazol-2-yl)methyl]amino)propan-2-ol dihydrochloride

  • MDL number MFCD18071274
  • Cas Number : 921146-02-3
  • Molecular Weight : 259.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7942

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-7942
Availability details for
Quantity
  • Availability
  • Add to Cart