COMH04238FDC Combi-Blocks, Inc.

5-([(4-Methylbenzyl)thio]methyl)-2-furoic acid

  • MDL number MFCD03834532
  • Cas Number : 696648-05-2
  • Molecular Weight : 262.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8075

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8075
Availability details for
Quantity
  • Availability
  • Add to Cart