COMH04238FF7 Combi-Blocks, Inc.

[2-Methyl-4-(2-morpholin-4-yl-2-oxoethoxy)phenyl]amine

  • MDL number MFCD05860754
  • Cas Number : 727674-88-6
  • Molecular Weight : 250.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8102

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8102
Availability details for
Quantity
  • Availability
  • Add to Cart