COMH04239016 Combi-Blocks, Inc.

1-(4-Chlorobenzyl)piperazin-2-one hydrochloride

  • MDL number MFCD13461791
  • Cas Number : 701208-33-5
  • Molecular Weight : 261.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8135

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8135
Availability details for
Quantity
  • Availability
  • Add to Cart