COMH0423906D Combi-Blocks, Inc.

3-(1-[(3-Fluorophenyl)methyl]-2,5-dioxoimidazolidin-4-yl)propanoic acid

  • MDL number MFCD29051090
  • Cas Number : 1955515-02-2
  • Molecular Weight : 280.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8222

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8222
Availability details for
Quantity
  • Availability
  • Add to Cart