COMH04239082 Combi-Blocks, Inc.

[(6,6-Dimethylbicyclo[3.1.1]hept-2-yl)methyl]amine

  • MDL number MFCD07809522
  • Cas Number : 38235-68-6
  • Molecular Weight : 153.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8244

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8244
Availability details for
Quantity
  • Availability
  • Add to Cart