COMH04239099 Combi-Blocks, Inc.

1-(1,3-Benzothiazol-2-yl)-5-oxopyrrolidine-3-carboxamide

  • MDL number MFCD16873296
  • Cas Number : 1239764-78-3
  • Molecular Weight : 261.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8268

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8268
Availability details for
Quantity
  • Availability
  • Add to Cart