COMH042390AA Combi-Blocks, Inc.

3-[2,5-Dioxo-1-(3,4,5-trifluorophenyl)imidazolidin-4-yl]propanoic acid

  • MDL number MFCD30723559
  • Cas Number : 2058426-46-1
  • Molecular Weight : 302.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8286

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8286
Availability details for
Quantity
  • Availability
  • Add to Cart