COMH042390B6 Combi-Blocks, Inc.

2-Bromo-n-methylpropanamide

  • MDL number MFCD13248679
  • Cas Number : 74538-22-0
  • Molecular Weight : 166.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8298

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8298
Availability details for
Quantity
  • Availability
  • Add to Cart