COMH042390BD Combi-Blocks, Inc.

Diethyl 5-amino-3-propylthiophene-2,4-dicarboxylate

  • MDL number MFCD03900731
  • Cas Number : 667437-85-6
  • Molecular Weight : 285.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8305

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8305
Availability details for
Quantity
  • Availability
  • Add to Cart