COMH042390C8 Combi-Blocks, Inc.

4-Amino-n-(3,4-dichlorophenyl)benzenesulfonamide

  • MDL number MFCD01016118
  • Cas Number : 34392-63-7
  • Molecular Weight : 317.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8316

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8316
Availability details for
Quantity
  • Availability
  • Add to Cart