COMH042390DE Combi-Blocks, Inc.

1-(1,3-Benzodioxol-5-ylmethyl)-1h-pyrazol-5-amine

  • MDL number MFCD07643287
  • Cas Number : 3528-55-0
  • Molecular Weight : 217.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8338

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8338
Availability details for
Quantity
  • Availability
  • Add to Cart