COMH042390E6 Combi-Blocks, Inc.

Ethyl 5-cyclopropyl-7-(difluoromethyl)pyrazolo[1,5-a]pyrimidine-3-carboxylate

  • MDL number MFCD03468748
  • Cas Number : 512825-94-4
  • Molecular Weight : 281.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8346

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8346
Availability details for
Quantity
  • Availability
  • Add to Cart