COMH042390EC Combi-Blocks, Inc.

2-(3,5-Dimethyl-1h-pyrazol-1-yl)phenol

  • MDL number MFCD01246360
  • Cas Number : 83430-99-3
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8352

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8352
Availability details for
Quantity
  • Availability
  • Add to Cart