COMH04239116 Combi-Blocks, Inc.

Ethyl [(methylamino)carbonothioyl]carbamate

  • MDL number MFCD13248748
  • Cas Number : 51863-43-5
  • Molecular Weight : 162.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8394

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8394
Availability details for
Quantity
  • Availability
  • Add to Cart