COMH0423911A Combi-Blocks, Inc.

4-(5,7-Dioxo-5,7-dihydro-6h-pyrrolo[3,4-b]pyrazin-6-yl)butanoic acid

  • MDL number MFCD20502376
  • Cas Number : 1351393-88-8
  • Molecular Weight : 235.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8398

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8398
Availability details for
Quantity
  • Availability
  • Add to Cart