COMH04239128 Combi-Blocks, Inc.

4-[(2-Fluorobenzyl)oxy]-3-methoxybenzoyl chloride

  • MDL number MFCD12197992
  • Cas Number : 926028-23-1
  • Molecular Weight : 294.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8412

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8412
Availability details for
Quantity
  • Availability
  • Add to Cart