COMH04239136 Combi-Blocks, Inc.

5-Ethyl-1H-indole-2,3-dione

  • MDL number MFCD01132047
  • Cas Number : 96202-56-1
  • Molecular Weight : 175.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8427

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8427
Availability details for
Quantity
  • Availability
  • Add to Cart