COMH04239178 Combi-Blocks, Inc.

4,5-Dimethyl-1h-1,3-benzodiazole

  • MDL number MFCD18807626
  • Cas Number : 69557-55-7
  • Molecular Weight : 146.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8494

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8494
Availability details for
Quantity
  • Availability
  • Add to Cart