COMH042391E3 Combi-Blocks, Inc.

1-(7-Methyl-2-[(2-methylbenzyl)thio][1,2,4]triazolo[1,5-a]pyrimidin-6-yl)ethanone

  • MDL number MFCD07469692
  • Cas Number : 879044-17-4
  • Molecular Weight : 312.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8601

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-8601
Availability details for
Quantity
  • Availability
  • Add to Cart